missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin H Mouse anti-Human, Mouse, Rat, CoraLite 594, Clone: 2C5G11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | Cyclin H |
|---|---|
| Clone | 2C5G11 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17781439
|
Proteintech
CL594-67065-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17689966
|
Proteintech
CL594-67065-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin H Monoclonal antibody apecifically detetects Cyclin H antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| Cyclin H | |
| Immunofluorescence | |
| CoraLite 594 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 902 | |
| Cyclin H fusion protein Ag28556 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 2C5G11 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC005280 | |
| CCNH | |
| IgG2a | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title