missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin H Mouse anti-Human, Mouse, Rat, CoraLite 594, Clone: 2C5G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67065-100UL
This item is not returnable.
View return policy
Description
Cyclin H Monoclonal antibody apecifically detetects Cyclin H antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| Cyclin H | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| CCNH | |
| Cyclin H fusion protein Ag28556 | |
| 100 μL | |
| 902 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG2a |
| Immunofluorescence | |
| 2C5G11 | |
| CoraLite 594 | |
| BC005280 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction