missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin E2 Recombinant Rabbit Monoclonal Antibody (23GB3535), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Cyclin E2 |
|---|---|
| Clone | 23GB3535 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088968
|
Invitrogen™
MA553624 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088907
|
Invitrogen™
MA553625 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin E2 Recombinant Monoclonal antibody specifically detects Cyclin E2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| Cyclin E2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O96020 | |
| 9134 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3535 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2 | |
| A synthetic peptide of human Cyclin E23 (111-133AA). | |
| Primary | |
| -20°C | |
| CCNE2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title