missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin E2 Recombinant Rabbit Monoclonal Antibody (23GB3535), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553625
This item is not returnable.
View return policy
Description
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
Specifications
| Cyclin E2 | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O96020 | |
| CCNE2 | |
| A synthetic peptide of human Cyclin E23 (111-133AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3535 | |
| Unconjugated | |
| CCNE2 | |
| Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9134 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction