missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D3 Mouse anti-Human, Clone: 1F2C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66357-1-IG-150UL
This item is not returnable.
View return policy
Description
The immunogen for the anti-h-Cyclin D3 was a recombit full length Cyclin D3 protein recognizing a 34 kD protein. Cyclin D3 is a G1 cyclin closedly related to Cyclins D1 and D2 phase progression. Like Cyclin D1 and Cyclin D2, it is activated by the same mechanisms and can phosphorylate pRB when associated with cdk4 and cdk6.Specifications
| Cyclin D3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30281 | |
| CCND3 | |
| Cyclin D3 Fusion Protein Ag22825 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1F2C6 | |
| Unconjugated | |
| CCND3 | |
| CCND3, Cyclin D3, G1/S specific cyclin D3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 896 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction