missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin A1 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

£155.00 - £386.00

Specifications

Antigen Cyclin A1
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17723544
View Documents
Invitrogen™
PA5122144
20 μL
£155.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17713544
GREENER_CHOICE
View Documents
Invitrogen™
PA5122143
100 μL
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.
Specifications

Specifications

Cyclin A1
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P78396, Q61456, Q6AY13
12427, 295052, 8900
Recombinant CCNA1 (Ser183-Tyr430) expressed in E. coli.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
CCNA1
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
CCNA1; CT146; cyclin A1; cyclin-A1; testicular tissue protein Li 34
CCNA1
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.