missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122144
This item is not returnable.
View return policy
Description
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.
Specifications
| Cyclin A1 | |
| Polyclonal | |
| Unconjugated | |
| CCNA1 | |
| CCNA1; CT146; cyclin A1; cyclin-A1; testicular tissue protein Li 34 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 12427, 295052, 8900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P78396, Q61456, Q6AY13 | |
| CCNA1 | |
| Recombinant CCNA1 (Ser183-Tyr430) expressed in E. coli. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction