missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyano-3-hydroxycinnamic acid, 98%

Product Code. 10307870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10307870 100 g 100g
10792581 5 g 5g
10533911 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307870 Supplier Thermo Scientific Acros Supplier No. 163091000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54673-07-3 | C10H7NO3 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54673-07-3
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00004203
InChI Key HPLNTJVXWMJLNJ-YWEYNIOJSA-N
Synonym 2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
PubChem CID 21338270
IUPAC Name (Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 230.0°C to 233.0°C
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H4CH=C(CN)CO2H
Quantity 100 g
Beilstein 10, 520
Formula Weight 189.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Cyano-3-hydroxycinnamic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • a-Cyano-3-hydroxycinnamic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.