missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Cy2 Bis NHS Esters
Shop All Cytiva Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
- Suitable for general labeling of amine residues.
- Large range of bulk packs available for greatest economy.
Specifications
Specifications
| Formulation | 100% Cy2 (Bis NHS ester |
| For Use With (Application) | Used for high-throughput screening (HTS) applications |
| Quantity | 50 mg |
| Product Type | Cy2 Bis NHS Ester |
| Kit Contents | Foil packs, each containing 5, 10 or 50mg dried dye |
| Molecular Weight (g/mol) | 896.95 |
| Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
| Label or Dye | Cy2 |
| Color | Yellow |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction