missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXXC5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58775-25ul
This item is not returnable.
View return policy
Description
CXXC5 Polyclonal specifically detects CXXC5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| CXXC5 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| CF5, CXXC finger 5, CXXC finger 5 protein, CXXC finger protein 5, CXXC-type zinc finger protein 5, HSPC195, Putative MAPK-activating protein PM08, Putative NF-kappa-B-activating protein 102, retinoid-inducible nuclear factor, RINF, WID, WT1-induced Inhibitor of Dishevelled | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| CXXC5 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ASLLANGHDLAAAMAVDKSNPTSKHKSGAVASLLSKAERATELAAEGQLTLQQFAQSTEMLKRVVQEHLPL | |
| 25 μL | |
| Signal Transduction | |
| 51523 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction