missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL5/ENA-78 Antibody (32B3) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£163.00 - £434.00
Specifications
| Antigen | CXCL5 |
|---|---|
| Clone | 32B3 |
| Concentration | LYOPH |
| Applications | ELISA, Blocking Assay, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18183655
|
Novus Biologicals
NBP2-22026-0.025MG |
0.025 mg |
£163.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18137471
|
Novus Biologicals
NBP2-22026 |
0.1 mg | |||||||
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Block/Neutralize.Specifications
| CXCL5 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5 | |
| CXCL5 | |
| Protein G purified |
| 32B3 | |
| ELISA, Blocking Assay, Western Blot | |
| Unconjugated | |
| Rat | |
| Cell Cycle and Replication | |
| P50228 | |
| 6374 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title