missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL5/ENA-78 Antibody (32B3) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22026-0.025MG
This item is not returnable.
View return policy
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Block/Neutralize.
Specifications
| CXCL5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, ELISA 1:1000-1:2000, Blocking/Neutralizing 50-200 molar excess | |
| P50228 | |
| CXCL5 | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse PlGF recombinant protein. Rat monoclonal anti Mouse CXCL5 P50228. | |
| 0.025 mg | |
| Cell Cycle and Replication | |
| 6374 | |
| Mouse | |
| Purified |
| ELISA, Blocking Assay, Western Blot | |
| 32B3 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction