missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL10 (IP-10) Monoclonal Antibody (4NY8UN), Alexa Fluor™ 647, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£146.00 - £387.00
Specifications
| Antigen | CXCL10 (IP-10) |
|---|---|
| Clone | 4NY8UN |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.Specifications
| CXCL10 (IP-10) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| 3627 | |
| IgG2b κ | |
| Affinity chromatography | |
| CXCL10 |
| 4NY8UN | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Human | |
| P02778 | |
| CXCL10 | |
| Primary | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title