missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£193.00 - £1415.00
Specifications
| Antigen | CXCL1/GRO alpha/KC/CINC-1 |
|---|---|
| Clone | 20326R |
| Dilution | Neutralization 0.600 - 9.00 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732597
![]() |
R&D Systems
MAB275R-SP |
25 μg |
£193.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18778488
![]() |
R&D Systems
MAB275R-100 |
100 μg |
£402.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18129190
|
R&D Systems
MAB275R-01M |
1 mg |
£1415.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human, Primate samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Neutralization 0.600 - 9.00 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| CXCL1 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 20326R | |
| Neutralization, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Primate | |
| 2919 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Detects human and primate CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title