missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB275R-01M
This item is not returnable.
View return policy
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human, Primate samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).
Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| CXCL1 | |
| E. coli-derived recombinant human CXCL1/GRO alpha/KC/CINC-1 | |
| 1 mg | |
| Primary | |
| Detects human and primate CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA, Flow Cytometry, Neutralization | |
| 20326R | |
| Neutralization 0.600 - 9.00 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2919 | |
| Primate, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction