missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CXADR Antibody (Med. Bio. Labs patent anti-CXADR) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | CXADR |
|---|---|
| Clone | Med. Bio. Labs patent anti-CXADR |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046800
|
Novus Biologicals™
NBP3-28411-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045489
|
Novus Biologicals™
NBP3-28411-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047268
|
Novus Biologicals™
NBP3-28411-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXADR Monoclonal antibody specifically detects CXADR in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| CXADR | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR | |
| CXADR | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Med. Bio. Labs patent anti-CXADR | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P78310 | |
| 1525 | |
| IgG2 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title