missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CXADR Antibody (Med. Bio. Labs patent anti-CXADR) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28411-100ug
This item is not returnable.
View return policy
Description
CXADR Monoclonal antibody specifically detects CXADR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXADR | |
| Monoclonal | |
| Unconjugated | |
| P78310 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Med. Bio. Labs patent anti-CXADR | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR | |
| CXADR | |
| 100 μg | |
| Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
| 1525 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction