missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CVT 313
GREENER_CHOICE

Product Code. 30000901
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000901

Brand: Tocris Bioscience™ 6174/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

cdk2 inhibitor

Cyclin-dependent kinase 2 (cdk2) inhibitor (reported IC50 values are 0.18 to 0.5 μM). Also inhibits cdk5 in sub-micromolar range (IC50 = 0.42 μM). Displays >7-fold selectivity for cdk2 and cdk5 over other cdks. Inhibits proliferation rat neonatal aortic smooth muscle cells and a range of tumor cells lines in vitro. Inhibits restenosis in a rat carotid artery injury model.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199986-75-9
Molecular Formula C20H28N6O3

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 400.47
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 400.48
Chemical Name or Material 2,2'-[[6-[[(4-Methoxyphenyl)methyl]amino]-9-(1-methylethyl)-9H-purin-2-yl]imino]bis[ethanol]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.