CAS RN 199986-75-9
CAS RN 199986-75-9
IUPAC Name:
2-[(2-hydroxyethyl)(6-{[(4-methoxyphenyl)methyl]amino}-9-(propan-2-yl)-9H-purin-2-yl)amino]ethan-1-ol
Synonyms:
2-[(2-hydroxyethyl)(9-isopropyl-6-{[(4-methoxyphenyl)methyl]amino}purin-2-yl)amino]ethanol
Molecular Weight (g/mol):
400.48
Molecular Formula:
C20H28N6O3
InChi Key:
NQVIIUBWMBHLOZ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CNC2=C3N=CN(C(C)C)C3=NC(=N2)N(CCO)CCO)C=C1
1
–
2
of
2
results
1
Tocris Bioscience™ CVT 313
CAS: 199986-75-9 Molecular Formula: C20H28N6O3
2
CVT-313, MedChemExpress
MedChemExpress CVT-313 (Cdk2 Inhibitor III) is a potent, selective, reversible, and ATP-competitive inhibitor of CDK2 with IC50 of 0.5 μM. CVT-313 inhibits CDC5L phosphorylation.