Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Di-repeats
Custom 5' Fluorescent Labeled/Unlabeled Pairs
Brand: Applied Biosystems™ 4304976
Code : NEW
Additional Details : Weight : 0.00100kg
Description
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.Specifications
Custom 5' Labeled/Unlabeled Di-Repeats, 10000 pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, Choice of Delivered Scales, These Products can be used in a Variety of Applications | |
-20°C | |
Fragment Analysis, Gene Expression Studies, Genotyping, PCR |
Custom 5' Labeled or Unlabeled Di-Repeats | |
10,000 pmol | |
Delivered either in Solution or Dried down |
For Research Use Only. Not for use in diagnostic procedures.