missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Di-repeats
Custom 5' Fluorescent Labeled/Unlabeled Pairs
£135.00 - £337.00
Specifications
Product Type | Custom 5' Labeled or Unlabeled Di-Repeats |
---|---|
Content And Storage | -20°C |
For Use With (Application) | Fragment Analysis, Gene Expression Studies, Genotyping, PCR |
Form | Delivered either in Solution or Dried down |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15537240
|
Applied Biosystems™
4304977 |
80,000 pmol |
£227.00
80000pmol |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15537280
|
Applied Biosystems™
4304978 |
300,000 pmol |
£337.00
300000pmol |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15507240
|
Applied Biosystems™
4304976 |
10,000 pmol |
£135.00
10000pmol |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.Specifications
Custom 5' Labeled or Unlabeled Di-Repeats | |
-20°C | |
Fragment Analysis, Gene Expression Studies, Genotyping, PCR | |
Delivered either in Solution or Dried down |
For Research Use Only. Not for use in diagnostic procedures.