missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cullin 4A Recombinant Rabbit Monoclonal Antibody (23GB790), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Cullin 4A |
|---|---|
| Clone | 23GB790 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088635
![]() |
Invitrogen™
MA553730 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089584
|
Invitrogen™
MA553731 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cullin 4A Recombinant Monoclonal antibody specifically detects Cullin 4A in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Cullin 4A | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13619 | |
| 8451 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB790 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853 | |
| A synthetic peptide of human beta Cullin4A (700-759AA). | |
| Primary | |
| -20°C | |
| CUL4A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title