missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cullin 4A Recombinant Rabbit Monoclonal Antibody (23GB790), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553731
This item is not returnable.
View return policy
Description
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
Specifications
| Cullin 4A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CUL4A | |
| 2810470J21Rik; AW495282; CUL 4A; Cul4a; CUL-4A; Cul4a protein; cullin 4A; cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753; RGD1563853 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8451 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB790 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13619 | |
| CUL4A | |
| A synthetic peptide of human beta Cullin4A (700-759AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction