missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cullin 3 Recombinant Rabbit Monoclonal Antibody (23GB1560), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Cullin 3 |
|---|---|
| Clone | 23GB1560 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089344
![]() |
Invitrogen™
MA552676 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088383
|
Invitrogen™
MA552677 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cullin 3 Recombinant Monoclonal antibody specifically detects Cullin 3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Cullin 3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| B5DF89, Q13618, Q9JLV5 | |
| 26554, 301555, 8452 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1560 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI467304; AW146203; Cul3; CUL-3; cullin 3; cullin-3; KIAA0617; mKIAA0617; OTTHUMP00000203839; OTTHUMP00000203840; PHA2E | |
| A synthesized peptide derived from human Cullin 3 (700-768AA). | |
| Primary | |
| -20°C | |
| CUL3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title