missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cullin 3 Recombinant Rabbit Monoclonal Antibody (23GB1560), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA552676
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Cullin 3 is a member of the family of human cullin genes (CUL 1, 2, 3, 4a, 4b, and 5) homologous to the S. cerevisiae cdc 53 gene. CUL 1 forms a complex with human p19 skp1 and F box protein p45 skp2. Together with cdc 53 (Known as Cul A), as three subunits, they form a ubiquitin ligase-controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL 1, CUL 2 does not associate with p19 skp1 and p45 skp2. Instead, CUL 2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL. The function of this association remains unknown.
Spécification
| Cullin 3 | |
| Recombinant Monoclonal | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B5DF89, Q13618, Q9JLV5 | |
| CUL3 | |
| A synthesized peptide derived from human Cullin 3 (700-768AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1560 | |
| Unconjugated | |
| CUL3 | |
| AI467304; AW146203; Cul3; CUL-3; cullin 3; cullin-3; KIAA0617; mKIAA0617; OTTHUMP00000203839; OTTHUMP00000203840; PHA2E | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26554, 301555, 8452 | |
| -20°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu