missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUGBP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £309.00
Specifications
| Antigen | CUGBP2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18268184
|
Novus Biologicals
NB600-204SS |
0.025 mL |
£154.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18291421
|
Novus Biologicals
NB600-204 |
0.1 mL |
£309.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CUGBP2 Polyclonal specifically detects CUGBP2 in Human, Mouse samples. It is validated for Western Blot.Specifications
| CUGBP2 | |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| 10659 | |
| C-terminus of human and mouse ETR-3 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 53 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein | |
| CELF2 | |
| IgG | |
| Affinity Purified | |
| This is specific for ETR-3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title