missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUGBP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-204
This item is not returnable.
View return policy
Description
CUGBP2 Polyclonal specifically detects CUGBP2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| CUGBP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CELF2 | |
| C-terminus of human and mouse ETR-3 | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for ETR-3. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000 | |
| BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein | |
| Rabbit | |
| 53 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Transcription Factors and Regulators | |
| 10659 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction