missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57682
This item is not returnable.
View return policy
Description
CUEDC1 Polyclonal specifically detects CUEDC1 in Human samples. It is validated for Western Blot.
Specifications
| CUEDC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CUE domain containing 1, CUE domain-containing protein 1, DKFZp547L163, FLJ20739 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 404093 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NWM3 | |
| CUEDC1 | |
| Synthetic peptides corresponding to CUEDC1(CUE domain containing 1) The peptide sequence was selected from the middle region of CUEDC1. Peptide sequence RNRDFLLALERDRLKYESQKSKSSSVAVGNDFGFSSPVPGTGDANPAVSE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Xenopus: 92%; Bovine: 92%; Rabbit: 92%; Zebrafish: 92%; Chicken: 91%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction