missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTR9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | CTR9 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18205052
|
Novus Biologicals
NBP2-57251 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643388
|
Novus Biologicals
NBP2-57251-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CTR9 Polyclonal specifically detects CTR9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| CTR9 | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 9646 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NILREHPNYVDCYLRLGAMARDKGNFYEASDWFKEALQINQDHPDAWSLIGNLHLAKQEWGPGQKKFERILKQPSTQSDTYSMLALGNVWLQT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Ctr9, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), KIAA0155SH2 domain-binding protein 1, p150TSP, RNA polymerase-associated protein CTR9 homolog, SH2 domain binding protein 1 (tetratricopeptide repeat containing), SH2BP1p150, TPR-containing, SH2-binding phosphoprotein, TSBP | |
| CTR9 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title