missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTP synthase Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CTP synthase |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612192
|
Novus Biologicals
NBP2-92435-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635582
|
Novus Biologicals
NBP2-92435-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CTP synthase Polyclonal antibody specifically detects CTP synthase in Human, Mouse samples. It is validated for Western BlotSpecifications
| CTP synthase | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1503 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CTP synthase, CTP synthase 1, CTP synthetase 1, cytidine 5-prime triphosphate synthetase, cytidine 5'-triphosphate synthetase, EC 6.3.4.2, UTP--ammonia ligase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 402-591 of human CTPS1 (NP_001896.2). MQLAVVEFSRNVLGWQDANSTEFDPTTSHPVVVDMPEHNPGQMGGTMRLGKRRTLFQTKNSVMRKLYGDADYLEERHRHRFEVNPVWKKCLEEQGLKFVGQDVEGERMEIVELEDHPFFVGVQYHPEFLSRPIKPSPPYFGLLLASVGRLSHYLQKGCRLSPRDTYSDRSGSSSPDSEITELKFPSINHD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title