missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTP synthase Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92435-0.02ml
This item is not returnable.
View return policy
Description
CTP synthase Polyclonal antibody specifically detects CTP synthase in Human, Mouse samples. It is validated for Western Blot
Specifications
| CTP synthase | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CTP synthase, CTP synthase 1, CTP synthetase 1, cytidine 5-prime triphosphate synthetase, cytidine 5'-triphosphate synthetase, EC 6.3.4.2, UTP--ammonia ligase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 402-591 of human CTPS1 (NP_001896.2). MQLAVVEFSRNVLGWQDANSTEFDPTTSHPVVVDMPEHNPGQMGGTMRLGKRRTLFQTKNSVMRKLYGDADYLEERHRHRFEVNPVWKKCLEEQGLKFVGQDVEGERMEIVELEDHPFFVGVQYHPEFLSRPIKPSPPYFGLLLASVGRLSHYLQKGCRLSPRDTYSDRSGSSSPDSEITELKFPSINHD | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 1503 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction