missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTGF/CCN2 Antibody (MM0038-6B13) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£158.00 - £440.00
Specifications
| Antigen | CTGF/CCN2 |
|---|---|
| Clone | MM0038-6B13 |
| Concentration | LYOPH |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240342
|
Novus Biologicals
NB110-61004-0.025MG |
0.025 mg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18063214
|
Novus Biologicals
NB110-61004 |
0.1 mg | |||||||
Description
CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CTGF/CCN2 | |
| LYOPH | |
| Immunohistochemistry, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| P29279 | |
| 1490 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| MM0038-6B13 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
| IgG1 | |
| Protein G purified | |
| Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title