missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTGF/CCN2 Antibody (MM0038-6B13) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-61004-0.025MG
This item is not returnable.
View return policy
Description
CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CTGF/CCN2 | |
| Monoclonal | |
| LYOPH | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| P29279 | |
| CTGF | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human Connective Tissue Growth Factor (CTGF). | |
| 0.025 mg | |
| Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 1490 | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, ELISA, Immunohistochemistry (Paraffin) | |
| MM0038-6B13 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Not yet tested in other species. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction