missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTGF/CCN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£153.00 - £409.00
Specifications
| Antigen | CTGF/CCN2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28562206) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18741284
|
Novus Biologicals
NB100-724SS |
0.025 mL |
£153.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18737923
|
Novus Biologicals
NB100-724 |
£409.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CTGF/CCN2 Polyclonal specifically detects CTGF/CCN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CTGF/CCN2 | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28562206) | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity | |
| PBS with 0.02% Sodium Azide | |
| CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
| CCN2 | |
| IgG | |
| Affinity Purified | |
| Human, mouse and rat. Predicted to react with porcine, bovine, chicken and sheep based on 100% sequence homology. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P29279 | |
| 1490 | |
| A synthetic peptide made to a C-terminal portion of human CTGF (between residues 299-349). [UniProt# P29279] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 38 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title