missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTGF/CCN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB100-724SS
This item is not returnable.
View return policy
Description
CTGF/CCN2 Polyclonal specifically detects CTGF/CCN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CTGF/CCN2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
| Rabbit | |
| 38 kDa | |
| 0.025 mL | |
| Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity | |
| 1490 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28562206) | |
| P29279 | |
| CCN2 | |
| A synthetic peptide made to a C-terminal portion of human CTGF (between residues 299-349). [UniProt# P29279] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse and rat. Predicted to react with porcine, bovine, chicken and sheep based on 100% sequence homology. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction