missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSPG4/NG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CSPG4/NG2 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856465
|
Proteintech
55027-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846465
|
Proteintech
55027-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to CSPG4.
NG2, a human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. NG2 represents an integral membrane chondroitin sulfate proteoglycan expressed by human maligt melanoma cells.Specifications
| CSPG4/NG2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q00657, Q6UVK1, Q8VHY0 | |
| 121021, 1464, 81651 | |
| Peptide | |
| Primary | |
| -20°C | |
| CSPG4 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CSPG4, HMW MAA, MCSP, MCSPG, MEL CSPG, MSK16, NG2 | |
| Cspg4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title