Learn More
CSPG4/NG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55027-1-AP-20UL
 View more versions of this product
Product Code. 16856465
  • £135.00 / 20µL
Description
The antibody is specific to CSPG4.
NG2, a human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. NG2 represents an integral membrane chondroitin sulfate proteoglycan expressed by human maligt melanoma cells.
Specifications
Specifications
CSPG4/NG2
Polyclonal
Unconjugated
CSPG4
CSPG4, HMW MAA, MCSP, MCSPG, MEL CSPG, MSK16, NG2
Rabbit
Antigen Affinity Chromatography
RUO
121021, 1464, 81651
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
Q00657, Q6UVK1, Q8VHY0
Cspg4
Peptide
20 μL
Primary
Mouse, Human, Rat