missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CSP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | CSP |
|---|---|
| Dilution | Western Blot, ELISA, Immunohistochemistry : 5-10 μg/mL |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066985
|
Novus Biologicals™
NBP3-30943-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067243
|
Novus Biologicals™
NBP3-30943-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSP Polyclonal antibody specifically detects CSP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western BlotSpecifications
| CSP | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| Phosphate buffered saline | |
| 80331 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot, ELISA, Immunohistochemistry : 5-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CSP, Cysteine string protein, cysteine string protein alpha, DKFZp434N1429, DKFZp761N1221, DnaJ (Hsp40) homolog, subfamily C, member 5, dnaJ homolog subfamily C member 5, DNAJC5A, FLJ00118, FLJ13070 | |
| Synthetic peptide derived from the human CSP protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title