missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CSP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30943-100ug
This item is not returnable.
View return policy
Description
CSP Polyclonal antibody specifically detects CSP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| CSP | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry : 5-10 μg/mL | |
| CSP, Cysteine string protein, cysteine string protein alpha, DKFZp434N1429, DKFZp761N1221, DnaJ (Hsp40) homolog, subfamily C, member 5, dnaJ homolog subfamily C member 5, DNAJC5A, FLJ00118, FLJ13070 | |
| Synthetic peptide derived from the human CSP protein. | |
| 100 μg | |
| Neuroscience | |
| 80331 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction