missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CSNK1A1 (CK1 alpha 1), GST Tag Recombinant Protein
Human CSNK1A1 (CK1 alpha 1), GST Tag Recombinant Protein
Brand: Invitrogen™ PR7361B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CKI proteins are monomeric, ranging from 25 to 55 kD, and are found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells.
Specifications
NP_001883, K164Q | |
66.4 kDa | |
CSNK1A1 (CK1 α 1), CK1 | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CK1 Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
100μg | |
CK1 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |