missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CSNK1A1 (CK1 alpha 1), GST Tag Recombinant Protein
Human CSNK1A1 (CK1 alpha 1), GST Tag Recombinant Protein
£1602.00 - £6900.00
Specifications
Accession Number | NP_001883, K164Q |
---|---|
Gene Alias | CSNK1A1 (CK1 α 1), CK1 |
KinaseFamily | CK1 Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428573
|
Invitrogen™
PR7361B |
100μg |
£1602.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10108782
|
Invitrogen™
PV4174 |
1mg |
£6900.00
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CKI proteins are monomeric, ranging from 25 to 55 kD, and are found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells.
Specifications
NP_001883, K164Q | |
CK1 Kinase Family | |
Recombinant | |
Baculovirus |
CSNK1A1 (CK1 α 1), CK1 | |
Human | |
GST Tag |