missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSK Mouse anti-Human, Mouse, Clone: 3B12C5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CSK |
|---|---|
| Clone | 3B12C5 |
| Concentration | 0.89 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16877695
|
Proteintech
66115-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16867695
|
Proteintech
66115-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.Specifications
| CSK | |
| 0.89 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C Src kinase, c src tyrosine kinase, CSK, Protein tyrosine kinase CYL, Tyrosine protein kinase CSK | |
| Csk | |
| IgG2b | |
| Protein A | |
| Antibody |
| 3B12C5 | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P41240, P41241 | |
| 12988, 1445 | |
| CSK Fusion Protein Ag12103 | |
| Primary | |
| -20°C | |
| CSK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title