missing translation for 'onlineSavingsMsg'
Learn More

CSK Mouse anti-Human, Mouse, Clone: 3B12C5, Proteintech

Product Code. 16877695 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16877695 20 μL 20µL
16867695 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16877695 Supplier Proteintech Supplier No. 661151IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.

Specifications

Antigen CSK
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Monoclonal
Clone 3B12C5
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSK
Gene Accession No. P41240, P41241
Gene Alias C Src kinase, c src tyrosine kinase, CSK, Protein tyrosine kinase CYL, Tyrosine protein kinase CSK
Gene Symbols Csk
Host Species Mouse
Immunogen CSK Fusion Protein Ag12103
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12988, 1445
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.