missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSK Antibody (CSK-04), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
£402.00
Specifications
| Antigen | CSK |
|---|---|
| Clone | CSK-04 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | HRP |
Description
CSK Monoclonal antibody specifically detects CSK in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| CSK | |
| Western Blot | |
| HRP | |
| Mouse | |
| Cell Cycle and Replication, Signal Transduction | |
| PBS | |
| 1445 | |
| IgG1 | |
| Protein A purified |
| CSK-04 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
| Bacterially expressed recombinant fragment of human Csk corresponding to amino acids 330-450. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title