missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSK Antibody (CSK-04), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-344H
This item is not returnable.
View return policy
Description
CSK Monoclonal antibody specifically detects CSK in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| CSK | |
| Monoclonal | |
| HRP | |
| C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK | |
| Bacterially expressed recombinant fragment of human Csk corresponding to amino acids 330-450. | |
| 0.1 mL | |
| Cell Cycle and Replication, Signal Transduction | |
| 1445 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| CSK-04 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction