missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSGLCAT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55085
This item is not returnable.
View return policy
Description
CSGLCAT Polyclonal specifically detects CSGLCAT in Human samples. It is validated for Western Blot.
Specifications
| CSGLCAT | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CHPF2 | |
| Synthetic peptides corresponding to CSGLCA-T The peptide sequence was selected from the N terminal of CSGLCA-T. Peptide sequence SLLRVSWIQGEGEDPCVEAVGERGGPQNPDSRARLDQSDEDFKPRIVPYY. | |
| Affinity purified | |
| RUO | |
| 54480 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Chondroitin glucuronyltransferase, chondroitin polymerizing factor 2KIAA1402chPF-2, Chondroitin synthase 3, chondroitin synthase-3, ChPF-2, CHSY3, ChSy-3chondroitin sulfate glucuronyltransferase, CSGlcA-T, CSGLCAT, EC 2.4.1.226, N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase | |
| Rabbit | |
| 18 kDa | |
| 100 μL | |
| Primary | |
| Canine: 79%; Porcine: 79%. | |
| Human, Pig, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction