missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSDE1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | CSDE1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227305
|
Novus Biologicals
NBP3-33278-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231789
|
Novus Biologicals
NBP3-33278-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSDE1 Monoclonal antibody specifically detects CSDE1 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| CSDE1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| cold shock domain containing E1, RNA-binding, cold shock domain-containing protein E1, D1S155EUNRRP5-1000E10.3, DKFZp779B0247, DKFZp779J1455, FLJ26882, KIAA0885, N-ras upstream gene protein, NRAS-related, NRU, Protein UNR, upstream of NRAS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human CSDE1 (NP_009089.4).,, Sequence:, MSFDPNLLHNNGHNGYPNGTSAALRETGVIEKLLTSYGFIQCSERQARLFFHCSQYNGNLQDLKVGDDVEFEVSSDRRTGKPIAVKLVKIKQEILPEERM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 7812 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title