missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Crystal Violet

Product Code. p-200104515
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16468448

Brand: MedChemExpress HYB0324A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Crystal violet (Basic Violet 3) is a triarylmethane dye. Crystal Violet (Gentian Violet) has antiviral effects against H1N1 and also has prominent bactericidal activities.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 10 g
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (245.11 mM) ∣ H2O : 5 mg/mL (12.26 mM; Need ultrasonic)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 407.98 g/mol
CAS 548-62-9
Synonym Basic Violet 3 ∣ Gentian Violet ∣ Methyl Violet 10B
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.