missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Crystal Violet

Product Code. 16468448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468448 10 g 10g
16460878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468448 Supplier MedChemExpress Supplier No. HYB0324A
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Crystal violet (Basic Violet 3) is a triarylmethane dye. Crystal Violet (Gentian Violet) has antiviral effects against H1N1 and also has prominent bactericidal activities.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 10 g
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (245.11 mM) ∣ H2O : 5 mg/mL (12.26 mM; Need ultrasonic)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 407.98 g/mol
CAS 548-62-9
Synonym Basic Violet 3 ∣ Gentian Violet ∣ Methyl Violet 10B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.