missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRX Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111078
This item is not returnable.
View return policy
Description
Immunogen sequence: PLPEAQRAGL VASGPSLTSA PYAMTYAPAS AFCSSPSAYG SPSSYFSGLD PYLSPMVPQL GGPALSPLSG PSVGPSLAQS PTSLSGQSYG AYSPVDSLEF K Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 97%.
The cone-rod homeobox-containing gene (CRX) encodes a transcription factor that coordinates the expression of several photoreceptor genes in the developing retina, including opsin and rhodopsin. Specifically, CRX binds the OTX motif (TAATCC/A) upstream from photoreceptor genes. The CRX gene is also expressed in the pinealocytes of the pineal gland and may regulate pineal circadian activity by controling the expression of melatonin synthesis genes. Furthermore, CRX(-) mice exhibit disruption of circadian rhythms. The human CRX gene maps to chromosome 19q13.3 within the region of the cone-rod dystrophy-2 locus (CORD2). Mutations in the CRX gene are implicated in the visual pathologies of CORD, Leber congenital amaurosis (LCA) and retinitis pigmentosa (RP). All characterized CRX gene mutations produce disease in heterozygotes although there is no known correlation between the pathologic phenotype and genetic mutation. Missense mutations of the CRX gene affect the homeobox domain, whereas frameshift mutations affect the OTX domain.
Specifications
| CRX | |
| Polyclonal | |
| Unconjugated | |
| Crx | |
| Cone rod homeobox; cone-rod homeobox; cone-rod homeobox containing; cone-rod homeobox protein; CORD 2; CORD2; CRD; CRX; Crx1; LCA 7; LCA7; orthodenticle homeobox 3; OTX; OTX 3; OTX3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1406 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43186 | |
| Crx | |
| Recombinant Protein corresonding to Human CRX. Recombinant protein control fragment (Product #RP-97575). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction